ENA388192055 Enamine

[1-(2-chlorophenyl)cyclopropyl]methanamine

  • MDL number MFCD07374421
  • Cas Number : 886365-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121871
Availability details for
Quantity
  • Availability
  • Add to Cart