ENA388192056 Enamine

ethyl 2-[3-(methoxymethyl)-1,2,4-oxadiazol-5-yl]acetate

  • MDL number MFCD21935331
  • Cas Number : 1423031-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121873
Availability details for
Quantity
  • Availability
  • Add to Cart