ENA388192057 Enamine

1,2,2a,3,4,5-hexahydroacenaphthylen-1-one

  • MDL number MFCD01851631
  • Cas Number : 63328-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121874
Availability details for
Quantity
  • Availability
  • Add to Cart