ENA388192063 Enamine

ethyl 5-(chlorosulfonyl)-1-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD09743910
  • Cas Number : 88398-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105628

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105628
Availability details for
Quantity
  • Availability
  • Add to Cart