ENA388192067 Enamine

1-(4,6-dimethylpyrimidin-2-yl)-5-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD10002155
  • Cas Number : 926016-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121910

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121910
Availability details for
Quantity
  • Availability
  • Add to Cart