ENA388192069 Enamine

2-chloro-6-(pyrrolidin-1-yl)benzaldehyde

  • MDL number MFCD11156344
  • Cas Number : 110405-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121912

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121912
Availability details for
Quantity
  • Availability
  • Add to Cart