ENA388192072 Enamine

3-chloro-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD20645782
  • Cas Number : 137343-52-3
  • Molecular Weight : 160.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-1-methyl-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C5H5ClN2O2/c1-8-2-3(5(9)10)4(6)7-8/h2H,1H3,(H,9,10)
Inchi Key OBOOPWDQTYAVEJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-121915

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-121915
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart