ENA388192073 Enamine

(2-methanesulfonyl-2-methylpropyl)(methyl)amine

  • MDL number MFCD16809864
  • Cas Number : 1249167-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121916
Availability details for
Quantity
  • Availability
  • Add to Cart