ENA388192075 Enamine

2-[(2-hydroxyethyl)sulfanyl]-5-nitrobenzoic acid

  • MDL number MFCD12144298
  • Cas Number : 124416-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121920
Availability details for
Quantity
  • Availability
  • Add to Cart