ENA388192078 Enamine

[2,2-dimethyl-3-(methylamino)propyl](methyl)[2-(pyrrolidin-1-yl)ethyl]amine

  • MDL number MFCD18293589
  • Cas Number : 1344270-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121924
Availability details for
Quantity
  • Availability
  • Add to Cart