ENA388192079 Enamine

(1S)-1-(3-chloro-2-fluorophenyl)ethan-1-amine

  • MDL number MFCD07772566
  • Cas Number : 1228561-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121925
Availability details for
Quantity
  • Availability
  • Add to Cart