ENA388192080 Enamine

(1R)-1-(3-chloro-2-fluorophenyl)ethan-1-amine

  • MDL number MFCD07772682
  • Cas Number : 1217449-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121927
Availability details for
Quantity
  • Availability
  • Add to Cart