ENA388192091 Enamine

methyl 1-bromocyclobutane-1-carboxylate

  • MDL number MFCD12922669
  • Cas Number : 51175-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121942
Availability details for
Quantity
  • Availability
  • Add to Cart