ENA388192093 Enamine

3-(4-phenoxyphenyl)prop-2-yn-1-ol

  • MDL number MFCD12521124
  • Cas Number : 500709-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42...44
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121944

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121944
Availability details for
Quantity
  • Availability
  • Add to Cart