ENA388192096 Enamine

2-(oxolan-3-yloxy)pyridin-4-amine

  • MDL number MFCD18364561
  • Cas Number : 1339933-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-121947
Availability details for
Quantity
  • Availability
  • Add to Cart