ENA388192102 Enamine

2-(4-cyano-5-methyl-1,2-oxazol-3-yl)acetic acid

  • MDL number MFCD18818530
  • Cas Number : 93276-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-121982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-121982
Availability details for
Quantity
  • Availability
  • Add to Cart