ENA388192109 Enamine

3-(ethylsulfanyl)benzoic acid

  • MDL number MFCD19666554
  • Cas Number : 5537-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95...97
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122001

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122001
Availability details for
Quantity
  • Availability
  • Add to Cart