ENA388192112 Enamine

3-(4-fluorophenyl)oxan-3-ol

  • MDL number MFCD19670464
  • Cas Number : 1338990-25-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122007
Availability details for
Quantity
  • Availability
  • Add to Cart