ENA388192123 Enamine

6-hydrazinylpyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD22570226
  • Cas Number : 1423031-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122217
Availability details for
Quantity
  • Availability
  • Add to Cart