ENA388192127 Enamine

(1-cyclohexylcyclohexyl)methanamine hydrochloride

  • MDL number MFCD22741291
  • Cas Number : 854707-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122237

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122237
Availability details for
Quantity
  • Availability
  • Add to Cart