ENA388192129 Enamine

2-(2,6-difluorophenyl)propanenitrile

  • MDL number MFCD11036486
  • Cas Number : 149680-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122239
Availability details for
Quantity
  • Availability
  • Add to Cart