ENA388192131 Enamine

2-{6-bromo-3H-imidazo[4,5-b]pyridin-2-yl}-4-chlorophenol

  • MDL number MFCD07339165
  • Cas Number : 863669-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122242

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122242
Availability details for
Quantity
  • Availability
  • Add to Cart