ENA388192139 Enamine

2-[3,5-dimethyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-4-yl]acetic acid

  • MDL number MFCD19611407
  • Cas Number : 1343894-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122252

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122252
Availability details for
Quantity
  • Availability
  • Add to Cart