ENA388192143 Enamine

1-(2-fluoro-5-methylphenyl)ethan-1-ol

  • MDL number MFCD17222662
  • Cas Number : 911218-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122257

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122257
Availability details for
Quantity
  • Availability
  • Add to Cart