ENA388192148 Enamine

1-[3-(thiophen-2-yl)-1,2,4-oxadiazol-5-yl]cyclohexan-1-amine hydrochloride

  • MDL number MFCD22741294
  • Cas Number : 1423033-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 224...226
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122262

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122262
Availability details for
Quantity
  • Availability
  • Add to Cart