ENA388192151 Enamine

2-[(4-aminopyridin-2-yl)oxy]ethan-1-ol

  • MDL number MFCD16084438
  • Cas Number : 1251131-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122266

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122266
Availability details for
Quantity
  • Availability
  • Add to Cart