ENA388192157 Enamine

2-(2,2-dimethyl-2,3-dihydro-1-benzofuran-7-yl)ethan-1-amine

  • MDL number MFCD08449369
  • Cas Number : 910387-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122274
Availability details for
Quantity
  • Availability
  • Add to Cart