ENA388192158 Enamine

5-cyclopropyl-1-ethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD14619945
  • Cas Number : 1250680-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122275

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122275
Availability details for
Quantity
  • Availability
  • Add to Cart