ENA388192165 Enamine

2-(4-chlorophenyl)-1-(4-ethoxyphenyl)ethan-1-one

  • MDL number MFCD12653596
  • Cas Number : 127727-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122283

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122283
Availability details for
Quantity
  • Availability
  • Add to Cart