ENA388192166 Enamine

6-methoxy-1,2-dihydronaphthalene

  • MDL number MFCD08544531
  • Cas Number : 60573-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122307
Availability details for
Quantity
  • Availability
  • Add to Cart