ENA388192173 Enamine

{[6-methyl-2-(piperidin-1-yl)pyrimidin-4-yl]methyl}(2-methylpropyl)amine

  • MDL number MFCD17384408
  • Cas Number : 1272301-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122315
Availability details for
Quantity
  • Availability
  • Add to Cart