ENA388192177 Enamine

2-methyl-1,3-benzothiazole-6-carbonitrile

  • MDL number MFCD16619173
  • Cas Number : 42474-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122323

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122323
Availability details for
Quantity
  • Availability
  • Add to Cart