ENA388192180 Enamine

2-(4-fluorophenyl)-2-hydroxypropanoic acid

  • MDL number MFCD11052520
  • Cas Number : 81170-13-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122326
Availability details for
Quantity
  • Availability
  • Add to Cart