ENA388192182 Enamine

1-ethyl-4-hydroxypyrrolidin-2-one

  • MDL number MFCD19665405
  • Cas Number : 68252-21-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122329
Availability details for
Quantity
  • Availability
  • Add to Cart