ENA388192185 Enamine

N-(3-chloro-4-fluorophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD11189064
  • Cas Number : 1039314-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122332
Availability details for
Quantity
  • Availability
  • Add to Cart