ENA388192186 Enamine

6-(4-hydroxypiperidine-1-carbonyl)-2-methyl-2,3,4,5-tetrahydropyridazin-3-one

  • MDL number MFCD12416430
  • Cas Number : 1156143-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-122335

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122335
Availability details for
Quantity
  • Availability
  • Add to Cart