ENA388192187 Enamine

1-[(3-methoxyphenyl)methyl]-1H-indol-6-amine

  • MDL number MFCD12665076
  • Cas Number : 1095600-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59...61
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122337

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122337
Availability details for
Quantity
  • Availability
  • Add to Cart