ENA388192189 Enamine

4-hydroxythiane-4-carboxylic acid

  • MDL number MFCD14628640
  • Cas Number : 50289-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122340

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122340
Availability details for
Quantity
  • Availability
  • Add to Cart