ENA388192190 Enamine

2-[(3-methoxypropyl)sulfanyl]-5-nitrobenzoic acid

  • MDL number MFCD19194498
  • Cas Number : 1274507-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122341
Availability details for
Quantity
  • Availability
  • Add to Cart