ENA388192193 Enamine

1-chloro-3,4-dimethyl-2,3-dihydro-1H-indene

  • MDL number MFCD18624487
  • Cas Number : 1410427-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122344
Availability details for
Quantity
  • Availability
  • Add to Cart