ENA388192195 Enamine

(1R)-1-(5-chloro-2-fluorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD16294082
  • Cas Number : 1332593-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122349

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122349
Availability details for
Quantity
  • Availability
  • Add to Cart