ENA388192197 Enamine

(1S)-1-(5-chloro-2-fluorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD22741298
  • Cas Number : 1423040-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122351

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122351
Availability details for
Quantity
  • Availability
  • Add to Cart