ENA388192230 Enamine

N-[1-(4-fluorophenyl)ethyl]hydroxylamine

  • MDL number MFCD11592159
  • Cas Number : 84271-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-122517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122517
Availability details for
Quantity
  • Availability
  • Add to Cart