ENA388192231 Enamine

2-(4-cyanophenyl)acetamide

  • MDL number MFCD18265396
  • Cas Number : 117753-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122518

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122518
Availability details for
Quantity
  • Availability
  • Add to Cart