ENA388192234 Enamine

2,2,2-trifluoro-N-(2-iodophenyl)acetamide

  • MDL number MFCD00547381
  • Cas Number : 143321-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122521

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122521
Availability details for
Quantity
  • Availability
  • Add to Cart