ENA388192238 Enamine

2-{[4-ethyl-5-(furan-2-yl)-4H-1,2,4-triazol-3-yl]sulfanyl}-1-(10H-phenothiazin-10-yl)ethan-1-one

  • MDL number MFCD04210733
  • Cas Number : 663212-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152...154
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122525

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122525
Availability details for
Quantity
  • Availability
  • Add to Cart