ENA388192241 Enamine

2-ethyl-5-methylphenol

  • MDL number MFCD08691687
  • Cas Number : 1687-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122559
Availability details for
Quantity
  • Availability
  • Add to Cart