ENA388192245 Enamine

5-(4-fluorophenoxymethyl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD22741302
  • Cas Number : 1423034-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154...156
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122563

Safety Information

msds https://www.enaminestore.com/catalog/EN300-122563
Availability details for
Quantity
  • Availability
  • Add to Cart