ENA388192248 Enamine

6-fluoro-5-nitroquinoline

  • MDL number MFCD11110425
  • Cas Number : 236092-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122567
Availability details for
Quantity
  • Availability
  • Add to Cart