ENA388192250 Enamine

[6-methyl-2-(piperidin-1-yl)pyrimidin-4-yl]methanol

  • MDL number MFCD17216994
  • Cas Number : 1342780-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-122569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-122569
Availability details for
Quantity
  • Availability
  • Add to Cart